Learn More
Medchemexpress LLC M-peg4-c6-phosphonic acid ethyl ester | 2028281-89-0 | C19H41O7P | 50 MG
SDP
Catalog No. 5000355321
Encompass_Preferred
M-PEG4-C6-phosphonic acid ethyl ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It facilitates targeted protein degradation by linking an E3 ubiquitin ligase ligand with a target protein ligand, exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Research use only
Catalog No. 50-003-55321 Supplier Medchemexpress LLC Supplier No. HY14131450MG
M-PEG4-C6-phosphonic acid ethyl ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It facilitates targeted protein degradation by linking an E3 ubiquitin ligase ligand with a target protein ligand, exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Research use only