Learn More
Medchemexpress LLC m-PEG4-CH2-methyl ester | 1920109-55-2 | MFCD26318564 | >98% | C12H24O6 | 25 MG
SDP
Catalog No. 5000352632
Encompass_Preferred
m-PEG4-CH2-methyl ester is a PROTAC linker that is based on PEG and Alkyl/ester. It can be utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. These molecules function by harnessing the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG- and Alkyl/ester-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • PROTACs selectively degrade target proteins by exploiting the ubiquitin-proteasome system.
Catalog No. 50-003-52632 Supplier Medchemexpress LLC Supplier No. 25MGHY141404
m-PEG4-CH2-methyl ester is a PROTAC linker that is based on PEG and Alkyl/ester. It can be utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. These molecules function by harnessing the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG- and Alkyl/ester-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • PROTACs selectively degrade target proteins by exploiting the ubiquitin-proteasome system.