Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG4-CH2COOH | 16024-66-1 | 99.9% | 1 G
SDP

Catalog No. 5000338733
Encompass_Preferred

This PEG-based PROTAC linker is used in the synthesis of PROTACs. It is intended for research applications.

  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Purity of 99.86%
  • Molecular formula C11H22O7
  • Molecular weight 266.29
  • Appears as a colorless to light yellow liquid
  • Specific storage conditions for pure form and in solvent
  • Soluble in various solvents for in vitro and in vivo studies
  • Relevant for cancer and cancer targeted therapy research

Catalog No. 50-003-38733 Supplier Medchemexpress LLC Supplier No. HY1304781G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This PEG-based PROTAC linker is used in the synthesis of PROTACs. It is intended for research applications.

  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Purity of 99.86%
  • Molecular formula C11H22O7
  • Molecular weight 266.29
  • Appears as a colorless to light yellow liquid
  • Specific storage conditions for pure form and in solvent
  • Soluble in various solvents for in vitro and in vivo studies
  • Relevant for cancer and cancer targeted therapy research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.