Learn More
Medchemexpress LLC m-PEG4-CH2COOH | 16024-66-1 | 99.9% | 250 MG
SDP
Catalog No. 5000341758
Encompass_Preferred
m-PEG4-CH2COOH is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs utilize two distinct ligands, connected by a linker, where one ligand binds to an E3 ubiquitin ligase and the other targets a specific protein. This mechanism exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-41758 Supplier Medchemexpress LLC Supplier No. 250MGHY130478
m-PEG4-CH2COOH is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs utilize two distinct ligands, connected by a linker, where one ligand binds to an E3 ubiquitin ligase and the other targets a specific protein. This mechanism exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only