Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG4-CH2COOH | 16024-66-1 | 99.9% | 5 G
SDP

Catalog No. 5000339965
Encompass_Preferred

m-PEG4-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.

  • Molecular weight: 266.29
  • Formula: C11H22O7
  • Appearance: Liquid (density: 1.125 g/cm³)
  • Color: Colorless to light yellow
  • Shipping at room temperature (in continental US)
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • In vitro solubility: DMSO: 100 mg/mL
  • In vivo solubility: ≥ 2.5 mg/mL in various protocols

Catalog No. 50-003-39965 Supplier Medchemexpress LLC Supplier No. HY1304785G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG4-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.

  • Molecular weight: 266.29
  • Formula: C11H22O7
  • Appearance: Liquid (density: 1.125 g/cm³)
  • Color: Colorless to light yellow
  • Shipping at room temperature (in continental US)
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • In vitro solubility: DMSO: 100 mg/mL
  • In vivo solubility: ≥ 2.5 mg/mL in various protocols

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.