Learn More
Medchemexpress LLC m-PEG4-Hydrazide | 1449390-68-4 | >98% | C10H22N2O5 | 25 MG
SDP
Catalog No. 5000352694
Encompass_Preferred
m-PEG4-Hydrazide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTAC linker
  • PEG-based
Catalog No. 50-003-52694 Supplier Medchemexpress LLC Supplier No. 25MGHY141402
m-PEG4-Hydrazide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTAC linker
  • PEG-based