Learn More
Medchemexpress LLC m-PEG4-Hydrazide | 1449390-68-4 | >98% | C10H22N2O5 | 250 MG
SDP
Catalog No. 5000357384
Encompass_Preferred
m-PEG4-Hydrazide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker, with one ligand for an E3 ubiquitin ligase and the other for the target protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only and not sold to patients.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • For research use only
Catalog No. 50-003-57384 Supplier Medchemexpress LLC Supplier No. 250MGHY141402
m-PEG4-Hydrazide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker, with one ligand for an E3 ubiquitin ligase and the other for the target protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only and not sold to patients.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • For research use only