Learn More
Medchemexpress LLC m-PEG4-Hydrazide | 1449390-68-4 | C10H22N2O5 | 1 G
SDP
Catalog No. 5000356739
Encompass_Preferred
m-PEG4-Hydrazide is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are chemical compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
Catalog No. 50-003-56739 Supplier Medchemexpress LLC Supplier No. HY1414021G
m-PEG4-Hydrazide is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are chemical compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins