Learn More
Medchemexpress LLC M-PEG4-hydrazide | 1449390-68-4 | C10H22N2O5 | 10MG
SDP
Catalog No. 5000260922
Encompass_Preferred
m-PEG4-hydrazide is a methoxy-terminated PEG4 linker containing a terminal hydrazide functional group. It is used as a spacer in PROTAC design and other bioconjugation workflows, enabling hydrazone formation with aldehydes while improving solubility and conformational flexibility. Supplied for research use; follow laboratory safety and handling guidance.
Catalog No. 50-002-60922 Supplier Medchemexpress LLC Supplier No. HY14140210MG
m-PEG4-hydrazide is a methoxy-terminated PEG4 linker containing a terminal hydrazide functional group. It is used as a spacer in PROTAC design and other bioconjugation workflows, enabling hydrazone formation with aldehydes while improving solubility and conformational flexibility. Supplied for research use; follow laboratory safety and handling guidance.