Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG4-MS | 130955-37-2 | ≥95.0% | 286.34 | C10H22O7S | 250 MG
SDP

Catalog No. 5000202504
Encompass_Preferred

m-PEG4-MS is a PEG-based linker used in the synthesis of PROTAC molecules and as a cleavable linker in antibody-drug conjugates (ADCs).

  • PEG-based linker for PROTACs and ADCs.
  • CAS number: 130955-37-2.
  • Purity: ≥95.0%.
  • Molecular formula: C10H22O7S.
  • Molecular weight: 286.34 g/mol.
  • Package size: 250 mg.

Catalog No. 50-002-02504 Supplier Medchemexpress LLC Supplier No. HY130457250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG4-MS is a PEG-based linker used in the synthesis of PROTAC molecules and as a cleavable linker in antibody-drug conjugates (ADCs).

  • PEG-based linker for PROTACs and ADCs.
  • CAS number: 130955-37-2.
  • Purity: ≥95.0%.
  • Molecular formula: C10H22O7S.
  • Molecular weight: 286.34 g/mol.
  • Package size: 250 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.