Learn More
Medchemexpress LLC M-PEG4-MS | 130955-37-2 | ≥95.0% | 286.34 | C10H22O7S | 500 MG
SDP
Catalog No. 5000203495
Encompass_Preferred
m-PEG4-MS is a polyethylene glycol (PEG)-based linker used in small-molecule conjugation chemistry. It is suitable as a PROTAC linker for assembling proteolysis-targeting chimeras and as a cleavable linker in antibody-drug conjugate (ADC) synthesis, enabling conjugation and controlled release in bioconjugation workflows.
  • Used in PROTAC synthesis
  • Functions as a cleavable ADC linker
  • PEG-based spacer for flexible linker design
  • High purity suitable for research use
  • Available as a 500 mg pack for preparative work
Catalog No. 50-002-03495 Supplier Medchemexpress LLC Supplier No. HY130457500MG
m-PEG4-MS is a polyethylene glycol (PEG)-based linker used in small-molecule conjugation chemistry. It is suitable as a PROTAC linker for assembling proteolysis-targeting chimeras and as a cleavable linker in antibody-drug conjugate (ADC) synthesis, enabling conjugation and controlled release in bioconjugation workflows.
  • Used in PROTAC synthesis
  • Functions as a cleavable ADC linker
  • PEG-based spacer for flexible linker design
  • High purity suitable for research use
  • Available as a 500 mg pack for preparative work