Learn More
Medchemexpress LLC M-PEG4-NH-DBCO | 2228857-36-9 | 95.0% | C28H34N2O6 | 100 MG
SDP
Catalog No. 5000353099
Encompass_Preferred
m-PEG4-NH-DBCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules that have Azide groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Used in synthesis of PROTACs
Catalog No. 50-003-53099 Supplier Medchemexpress LLC Supplier No. HY140315100MG
m-PEG4-NH-DBCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules that have Azide groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Used in synthesis of PROTACs