Learn More
Medchemexpress LLC m-PEG4-NH-DBCO | 2228857-36-9 | MFCD31654033 | ≥95.0% | C₂₈H₃₄N₂O₆ | 50 MG
SDP
Catalog No. 5000353035
Encompass_Preferred
m-PEG4-NH-DBCO is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent, enabling strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. PROTACs utilize the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Features a DBCO group
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Utilizes the ubiquitin-proteasome system
  • Achieves selective degradation of target proteins
  • For research use only
Catalog No. 50-003-53035 Supplier Medchemexpress LLC Supplier No. HY14031550MG
m-PEG4-NH-DBCO is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent, enabling strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. PROTACs utilize the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Features a DBCO group
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Utilizes the ubiquitin-proteasome system
  • Achieves selective degradation of target proteins
  • For research use only