Learn More
Medchemexpress LLC M-PEG4-PFP ester | 2952895-31-5 | ≥98.0% | 402.31 | 100 MG
SDP
Catalog No. 5000348634
Encompass_Preferred
m-PEG4-PFP ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules are engineered to contain two distinct ligands, one binding to an E3 ubiquitin ligase and the other targeting a specific protein. This innovative design allows PROTACs to leverage the cell's ubiquitin-proteasome system for the selective degradation of target proteins.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-48634 Supplier Medchemexpress LLC Supplier No. HY138382100MG
m-PEG4-PFP ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules are engineered to contain two distinct ligands, one binding to an E3 ubiquitin ligase and the other targeting a specific protein. This innovative design allows PROTACs to leverage the cell's ubiquitin-proteasome system for the selective degradation of target proteins.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins