Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-PEG4-PFP ester | 2952895-31-5 | 98.0% | 402.31 | 25 MG
SDP

Catalog No. 5000350474
Encompass_Preferred

M-PEG4-PFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.

  • Molecular weight: 402.31
  • Formula: C16H19F5O6
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Solubility in DMSO: ≥ 100 mg/mL (248.56 mM)

Catalog No. 50-003-50474 Supplier Medchemexpress LLC Supplier No. 25MGHY138382
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M-PEG4-PFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.

  • Molecular weight: 402.31
  • Formula: C16H19F5O6
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Solubility in DMSO: ≥ 100 mg/mL (248.56 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.