Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG4-phosphonic acid | 1872433-62-9 | 99.36% | C9H21O7P | 50 MG
SDP

Catalog No. 5000356303
Encompass_Preferred

m-PEG4-phosphonic acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Molecular weight: 272.23
  • Formula: C9H21O7P
  • CAS number: 1872433-62-9
  • Appearance: Oil
  • Color: Brown to reddish brown
  • Solubility: 25 mg/mL (91.83 mM) in DMSO
  • Purity: 99.36%
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-56303 Supplier Medchemexpress LLC Supplier No. HY14130750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG4-phosphonic acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Molecular weight: 272.23
  • Formula: C9H21O7P
  • CAS number: 1872433-62-9
  • Appearance: Oil
  • Color: Brown to reddish brown
  • Solubility: 25 mg/mL (91.83 mM) in DMSO
  • Purity: 99.36%
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.