Learn More
Medchemexpress LLC m-PEG4-phosphonic acid | 1872433-62-9 | 99.4% | C9H21O7P | 100 MG
SDP
Catalog No. 5000355317
Encompass_Preferred
m-PEG4-phosphonic acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds comprising two different ligands connected by a linker. One ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein. This mechanism allows PROTACs to utilize the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Key component in the development of targeted therapy drugs
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-55317 Supplier Medchemexpress LLC Supplier No. HY141307100MG
m-PEG4-phosphonic acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds comprising two different ligands connected by a linker. One ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein. This mechanism allows PROTACs to utilize the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Key component in the development of targeted therapy drugs
  • Facilitates selective degradation of target proteins