Learn More
Medchemexpress LLC m-PEG4-phosphonic acid ethyl ester | 1872433-73-2 | C13H29O7P | 100 MG
SDP
Catalog No. 5000355320
Encompass_Preferred
m-PEG4-phosphonic acid ethyl ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This product is for research use only and not for sale to patients.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-55320 Supplier Medchemexpress LLC Supplier No. HY141311100MG
m-PEG4-phosphonic acid ethyl ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This product is for research use only and not for sale to patients.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins