Learn More
Medchemexpress LLC m-PEG4-sulfonic acid | 787524-78-1 | 97.0% | 272.32 | 50 MG
SDP
Catalog No. 5000349481
Encompass_Preferred
m-PEG4-sulfonic acid is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other for the target protein. PROTACs operate by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-49481 Supplier Medchemexpress LLC Supplier No. HY14017050MG
m-PEG4-sulfonic acid is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other for the target protein. PROTACs operate by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only