Learn More
Medchemexpress LLC m-PEG5-acid | 81836-43-3 | 95.0% | 280.31 g/mol | C12H24O7 | 5 G
SDP
Catalog No. 5000209715
Encompass_Preferred
m-PEG5-acid is a methoxy-terminated polyethylene glycol linker (five ethylene glycol units) with a terminal carboxylic acid that provides a short, flexible spacer for PROTAC and bioconjugation chemistry.
  • provides a short, flexible peg spacer for linker design
  • contains a terminal carboxylic acid for coupling reactions
  • methoxy-terminated to cap one end and prevent further reaction
  • useful in synthesis of PROTACs and other bioconjugates
  • available in small laboratory pack sizes for research use
Catalog No. 50-002-09715 Supplier Medchemexpress LLC Supplier No. HYW0401955G
m-PEG5-acid is a methoxy-terminated polyethylene glycol linker (five ethylene glycol units) with a terminal carboxylic acid that provides a short, flexible spacer for PROTAC and bioconjugation chemistry.
  • provides a short, flexible peg spacer for linker design
  • contains a terminal carboxylic acid for coupling reactions
  • methoxy-terminated to cap one end and prevent further reaction
  • useful in synthesis of PROTACs and other bioconjugates
  • available in small laboratory pack sizes for research use