Learn More
Medchemexpress LLC M-PEG5-acid | 81836-43-3 | MFCD20926398 | 95.0% | 280.31 g/mol | C12H24O7 | 1 G
SDP
Catalog No. 5000208924
Encompass_Preferred
m-PEG5-acid is a monomethylated polyethylene glycol (PEG) linker terminated with a carboxylic acid group, used as a short spacer in the synthesis of PROTACs and other bioconjugates. It provides a PEG5 (approximately five ethylene oxide units) spacer with a reactive carboxylic acid for standard coupling chemistries.
  • Short monomethylated PEG5 spacer with terminal carboxylic acid.
  • Useful as a linker in PROTAC and conjugate synthesis.
  • Molecular weight ~280.31 g/mol and formula C12H24O7.
  • Typical purity around 95.0%.
  • Light yellow to light brown liquid; density ~1.107 g/cm3.
  • Store sealed, away from moisture; use cold storage for solutions.
Catalog No. 50-002-08924 Supplier Medchemexpress LLC Supplier No. HYW0401951G
m-PEG5-acid is a monomethylated polyethylene glycol (PEG) linker terminated with a carboxylic acid group, used as a short spacer in the synthesis of PROTACs and other bioconjugates. It provides a PEG5 (approximately five ethylene oxide units) spacer with a reactive carboxylic acid for standard coupling chemistries.
  • Short monomethylated PEG5 spacer with terminal carboxylic acid.
  • Useful as a linker in PROTAC and conjugate synthesis.
  • Molecular weight ~280.31 g/mol and formula C12H24O7.
  • Typical purity around 95.0%.
  • Light yellow to light brown liquid; density ~1.107 g/cm3.
  • Store sealed, away from moisture; use cold storage for solutions.