Learn More
Medchemexpress LLC M-PEG5-azide | 1202681-04-6 | 1 G
SDP
Catalog No. 5000340509
Encompass_Preferred
M-PEG5-azide is a PEG-based PROTAC linker crucial for the synthesis of PROTACs. This click chemistry reagent contains an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAC) with alkyne-containing molecules. It also facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules that have DBCO or BCN groups.
  • Peg-based protac linker
  • Used in protac synthesis
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC)
  • Also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-40509 Supplier Medchemexpress LLC Supplier No. HY1301681G
M-PEG5-azide is a PEG-based PROTAC linker crucial for the synthesis of PROTACs. This click chemistry reagent contains an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAC) with alkyne-containing molecules. It also facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules that have DBCO or BCN groups.
  • Peg-based protac linker
  • Used in protac synthesis
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC)
  • Also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)