Learn More
Medchemexpress LLC M-PEG5-azide | 1202681-04-6 | 50 MG
SDP
Catalog No. 5000338562
Encompass_Preferred
M-PEG5-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent features an azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Functions as a PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Acts as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN-containing molecules
Catalog No. 50-003-38562 Supplier Medchemexpress LLC Supplier No. HY13016850MG
M-PEG5-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent features an azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Functions as a PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Acts as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN-containing molecules