Learn More
Medchemexpress LLC M-PEG5-CH2CH2COOH | 1347750-72-4 | MFCD20926383 | ≥95.0% | 324.37 g/mol | C14H28O8 | 250 MG
SDP
Catalog No. 5000209406
Encompass_Preferred
m-PEG5-CH2CH2COOH is a methoxy-capped PEG5 propionic acid linker used as a PEG spacer in the synthesis of PROTACs and other bioconjugates. Supplied as a purified solid reagent, it is intended for research use in conjugation and organic synthesis.
  • High purity suitable for research applications.
  • PEG5 spacer provides flexible linkage and improved solubility.
  • Carboxylic acid terminus enables amide or ester coupling reactions.
  • Available in multiple pack sizes for scale flexibility.
  • Provided with datasheet, SDS, and COA for quality assurance.
Catalog No. 50-002-09406 Supplier Medchemexpress LLC Supplier No. 250MGHYW043840
m-PEG5-CH2CH2COOH is a methoxy-capped PEG5 propionic acid linker used as a PEG spacer in the synthesis of PROTACs and other bioconjugates. Supplied as a purified solid reagent, it is intended for research use in conjugation and organic synthesis.
  • High purity suitable for research applications.
  • PEG5 spacer provides flexible linkage and improved solubility.
  • Carboxylic acid terminus enables amide or ester coupling reactions.
  • Available in multiple pack sizes for scale flexibility.
  • Provided with datasheet, SDS, and COA for quality assurance.