Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG5-NH2 | 5498-83-9 | 99.0% | C11H25NO5 | 250 MG
SDP

Catalog No. 5000352700
Encompass_Preferred

m-PEG5-NH2 is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules consisting of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. These molecules function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only and not sold to patients.

  • Purity: 99.02%
  • Molecular weight: 251.32
  • Formula: C11H25NO5
  • CAS number: 5498-83-9
  • Appearance: Liquid (density: 1.023±0.06 g/cm³)
  • Color: Colorless to light yellow
  • SMILES: COCCOCCOCCOCCOCCN
  • Leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

Catalog No. 50-003-52700 Supplier Medchemexpress LLC Supplier No. 250MGHY140225
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG5-NH2 is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules consisting of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. These molecules function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only and not sold to patients.

  • Purity: 99.02%
  • Molecular weight: 251.32
  • Formula: C11H25NO5
  • CAS number: 5498-83-9
  • Appearance: Liquid (density: 1.023±0.06 g/cm³)
  • Color: Colorless to light yellow
  • SMILES: COCCOCCOCCOCCOCCN
  • Leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.