Learn More
Medchemexpress LLC HY-133272 100mg Medchemexpress, m-PEG5-SH CAS:524030-00-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY133272100MG
Encompass_Preferred
Medchemexpress, HY-133272 100mg m-PEG5-SH CAS:524030-00-0 m-PEG5-SH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1335