Learn More
Medchemexpress LLC m-PEG5-succinimidyl carbonate | 1058691-00-1 | 98.0% | 393.39 g/mol | C16H27NO10 | 25 MG
SDP
Catalog No. 5000202173
Encompass_Preferred
m-PEG5-succinimidyl carbonate is a monomethoxy polyethylene glycol (PEG) linker with five ethylene glycol units terminated as a succinimidyl carbonate active ester. It is supplied as a reactive linker for conjugation to primary amines in bioconjugation workflows.
  • Non-cleavable 5-unit PEG spacer for conjugation.
  • Activated succinimidyl carbonate for efficient amine coupling.
  • Suitable for ADC and PROTAC synthesis applications.
  • Appearance: colorless to light yellow liquid; supplied in small mg quantities for research use.
Catalog No. 50-002-02173 Supplier Medchemexpress LLC Supplier No. HY13015025MG
m-PEG5-succinimidyl carbonate is a monomethoxy polyethylene glycol (PEG) linker with five ethylene glycol units terminated as a succinimidyl carbonate active ester. It is supplied as a reactive linker for conjugation to primary amines in bioconjugation workflows.
  • Non-cleavable 5-unit PEG spacer for conjugation.
  • Activated succinimidyl carbonate for efficient amine coupling.
  • Suitable for ADC and PROTAC synthesis applications.
  • Appearance: colorless to light yellow liquid; supplied in small mg quantities for research use.