Learn More
Medchemexpress LLC M-PEG6-2-methylacrylate | 90784-86-4 | 250 MG
SDP
Catalog No. 5000342801
Encompass_Preferred
m-PEG6-2-methylacrylate is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs employ the intracellular ubiquitin-proteasome system for the selective degradation of target proteins, functioning by connecting ligands for an E3 ubiquitin ligase and a target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-42801 Supplier Medchemexpress LLC Supplier No. 250MGHY130548
m-PEG6-2-methylacrylate is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs employ the intracellular ubiquitin-proteasome system for the selective degradation of target proteins, functioning by connecting ligands for an E3 ubiquitin ligase and a target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins