Learn More
Medchemexpress LLC m-PEG6-amino-Mal | 1644231-07-1 | 100 MG
SDP
Catalog No. 5000340504
Encompass_Preferred
m-PEG6-amino-Mal is a chemical substance intended for use as a laboratory chemical and in the manufacture of other substances. It is stable when stored under recommended conditions.
  • Avoid inhalation and contact with eyes and skin.
  • Use in areas with appropriate exhaust ventilation.
  • Keep container tightly sealed in a cool, well-ventilated area.
  • Store away from direct sunlight and sources of ignition.
  • Recommended storage for powder is -20°C for 3 years or 4°C for 2 years.
  • Recommended storage in solvent is -80°C for 6 months.
  • For shipping less than 2 weeks, store at -20°C for 1 month.
Catalog No. 50-003-40504 Supplier Medchemexpress LLC Supplier No. HY130141100MG
m-PEG6-amino-Mal is a chemical substance intended for use as a laboratory chemical and in the manufacture of other substances. It is stable when stored under recommended conditions.
  • Avoid inhalation and contact with eyes and skin.
  • Use in areas with appropriate exhaust ventilation.
  • Keep container tightly sealed in a cool, well-ventilated area.
  • Store away from direct sunlight and sources of ignition.
  • Recommended storage for powder is -20°C for 3 years or 4°C for 2 years.
  • Recommended storage in solvent is -80°C for 6 months.
  • For shipping less than 2 weeks, store at -20°C for 1 month.