Learn More
Medchemexpress LLC m-PEG6-azide 250mg | 1043884-49-6 | 321.37 g/mol | C13H27N3O6 | 250 MG
SDP
Catalog No. 5000230081
Encompass_Preferred
m-PEG6-azide is a six-unit monomethyl polyethylene glycol linker bearing a terminal azide group. It is used as a PEG spacer in bioconjugation and antibody-drug conjugate linker synthesis, where the azide enables click chemistry (for example, CuAAC) and other azide-based conjugations.
  • Six-unit monomethyl PEG with a terminal azide functional group.
  • Enables click chemistry and azide-based bioconjugation.
  • Used as a PEG spacer in ADC linker synthesis and general bioconjugation.
  • High purity (98.0%) for consistent conjugation performance.
  • Colorless to light yellow liquid; store at -20°C (pure) for long-term stability.
  • Supplied as a 250 mg pack suitable for small-scale synthesis.
Catalog No. 50-002-30081 Supplier Medchemexpress LLC Supplier No. HY115374250MG
m-PEG6-azide is a six-unit monomethyl polyethylene glycol linker bearing a terminal azide group. It is used as a PEG spacer in bioconjugation and antibody-drug conjugate linker synthesis, where the azide enables click chemistry (for example, CuAAC) and other azide-based conjugations.
  • Six-unit monomethyl PEG with a terminal azide functional group.
  • Enables click chemistry and azide-based bioconjugation.
  • Used as a PEG spacer in ADC linker synthesis and general bioconjugation.
  • High purity (98.0%) for consistent conjugation performance.
  • Colorless to light yellow liquid; store at -20°C (pure) for long-term stability.
  • Supplied as a 250 mg pack suitable for small-scale synthesis.