Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG6-CH2CH2CHO | 1058691-77-2 | >98.0% | 352.42 g/mol | 250 MG
SDP

Catalog No. 5000202908
Encompass_Preferred

m-PEG6-CH2CH2CHO is a PEG-based linker reagent with a terminal aldehyde group used for conjugation in the synthesis of antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs). It is supplied as a research-grade reagent.

  • Non-cleavable ADC linker used for bioconjugation
  • Aldehyde functional group for covalent conjugation
  • Molecular weight 352.42 g/mol
  • CAS number 1058691-77-2
  • Purity >98.0%
  • Typical pack size 250 MG
  • Solubility approximately 10 mM in DMSO
  • For research use only; not for clinical or diagnostic use

Catalog No. 50-002-02908 Supplier Medchemexpress LLC Supplier No. HYW035376250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG6-CH2CH2CHO is a PEG-based linker reagent with a terminal aldehyde group used for conjugation in the synthesis of antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs). It is supplied as a research-grade reagent.

  • Non-cleavable ADC linker used for bioconjugation
  • Aldehyde functional group for covalent conjugation
  • Molecular weight 352.42 g/mol
  • CAS number 1058691-77-2
  • Purity >98.0%
  • Typical pack size 250 MG
  • Solubility approximately 10 mM in DMSO
  • For research use only; not for clinical or diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.