Learn More
Medchemexpress LLC m-PEG6-CH2CH2COOH | 874208-91-0 | >=95.0% | 368.42 g/mol | C16H32O9 | 500 MG
SDP
Catalog No. 5000208774
Encompass_Preferred
m-PEG6-CH2CH2COOH is a polyethylene glycol (PEG)-based PROTAC linker terminated with a carboxylic acid. It provides a six-unit PEG spacer for flexible linkage in conjugation chemistry and is supplied as a solid for synthetic use. Typical quality: ≥95.0% purity; molecular weight 368.42 g/mol; formula C16H32O9; CAS 874208-91-0.
  • Peg6 spacer with terminal carboxylic acid for conjugation.
  • Suitable for use as a PROTAC or linker building block in medicinal chemistry.
  • Available in multiple pack sizes, including 500 MG.
  • High purity for synthetic applications (≥95.0%).
  • Solid form with defined molecular weight (368.42 g/mol).
Catalog No. 50-002-08774 Supplier Medchemexpress LLC Supplier No. HYW040239500MG
m-PEG6-CH2CH2COOH is a polyethylene glycol (PEG)-based PROTAC linker terminated with a carboxylic acid. It provides a six-unit PEG spacer for flexible linkage in conjugation chemistry and is supplied as a solid for synthetic use. Typical quality: ≥95.0% purity; molecular weight 368.42 g/mol; formula C16H32O9; CAS 874208-91-0.
  • Peg6 spacer with terminal carboxylic acid for conjugation.
  • Suitable for use as a PROTAC or linker building block in medicinal chemistry.
  • Available in multiple pack sizes, including 500 MG.
  • High purity for synthetic applications (≥95.0%).
  • Solid form with defined molecular weight (368.42 g/mol).