Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG6-NHS ester | 1449390-12-8 | ≥97.0% | 421.44 g·mol⁻¹ | C18H31NO10 | 50 MG
SDP

Catalog No. 5000203672
Encompass_Preferred

m-PEG6-NHS ester is a non-cleavable 6-unit polyethylene glycol (PEG) linker terminating in an activated N-hydroxysuccinimide (NHS) ester; it is used for amide coupling to amines in the synthesis of antibody-drug conjugates and PROTACs.

  • Purity: ≥97.0%.
  • Molecular weight: 421.44 g·mol⁻¹.
  • Chemical formula: C18H31NO10.
  • CAS number: 1449390-12-8.
  • Form: liquid, colorless to light yellow; density ~1.21 g/cm³.
  • Storage: pure form -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
  • Applications: non-cleavable PEG6 linker for ADC synthesis; PEG/alkyl/ether-based PROTAC linker.
  • Packaging: commonly available in 100 MG, 250 MG, 1 G, and 5 G; larger quantities available by quote; a 50 MG pack is indicated in supplier data but on the manufacturer page.

Catalog No. 50-002-03672 Supplier Medchemexpress LLC Supplier No. HY13306650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG6-NHS ester is a non-cleavable 6-unit polyethylene glycol (PEG) linker terminating in an activated N-hydroxysuccinimide (NHS) ester; it is used for amide coupling to amines in the synthesis of antibody-drug conjugates and PROTACs.

  • Purity: ≥97.0%.
  • Molecular weight: 421.44 g·mol⁻¹.
  • Chemical formula: C18H31NO10.
  • CAS number: 1449390-12-8.
  • Form: liquid, colorless to light yellow; density ~1.21 g/cm³.
  • Storage: pure form -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
  • Applications: non-cleavable PEG6 linker for ADC synthesis; PEG/alkyl/ether-based PROTAC linker.
  • Packaging: commonly available in 100 MG, 250 MG, 1 G, and 5 G; larger quantities available by quote; a 50 MG pack is indicated in supplier data but on the manufacturer page.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.