Learn More
Medchemexpress LLC m-PEG6-thiol | 441771-60-4 | >98% | C13H28O6S | 250 MG
SDP
Catalog No. 5000352852
Encompass_Preferred
m-PEG6-thiol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs operate by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They consist of two different ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52852 Supplier Medchemexpress LLC Supplier No. 250MGHY141329
m-PEG6-thiol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs operate by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They consist of two different ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins