Learn More
Medchemexpress LLC M-peg6-thiol | 441771-60-4 | C13H28O6S | 100 MG
SDP
Catalog No. 5000355322
Encompass_Preferred
M-PEG6-thiol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for a specific target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to facilitate selective degradation of target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Targets specific proteins for degradation
  • Exploits the ubiquitin-proteasome system
Catalog No. 50-003-55322 Supplier Medchemexpress LLC Supplier No. HY141329100MG
M-PEG6-thiol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for a specific target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to facilitate selective degradation of target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Targets specific proteins for degradation
  • Exploits the ubiquitin-proteasome system