Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG7-amine | 170572-38-0 | MFCD13184965 | 339.43 g/mol | C15H33NO7 | 250 MG
SDP

Catalog No. 5000204258
Encompass_Preferred

m-PEG7-amine is a monofunctional polyethylene glycol (PEG) linker bearing a terminal amine. It is used as a hydrophilic spacer in conjugation chemistry, including PROTAC and antibody-drug conjugate (ADC) synthesis, to improve solubility and provide a reactive handle for bioconjugation.

  • Used as a PEG linker in PROTAC and ADC synthesis.
  • Contains a terminal primary amine for conjugation to activated esters and carbonyls.
  • Molecular weight approximately 339.43, formula C15H33NO7.
  • Soluble in DMSO (100 mg/mL; ultrasonic assistance may be required).
  • Compatible with common in vivo co-solvent formulations at ≥2.5 mg/mL.
  • Store protected from light at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-04258 Supplier Medchemexpress LLC Supplier No. HY120237250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG7-amine is a monofunctional polyethylene glycol (PEG) linker bearing a terminal amine. It is used as a hydrophilic spacer in conjugation chemistry, including PROTAC and antibody-drug conjugate (ADC) synthesis, to improve solubility and provide a reactive handle for bioconjugation.

  • Used as a PEG linker in PROTAC and ADC synthesis.
  • Contains a terminal primary amine for conjugation to activated esters and carbonyls.
  • Molecular weight approximately 339.43, formula C15H33NO7.
  • Soluble in DMSO (100 mg/mL; ultrasonic assistance may be required).
  • Compatible with common in vivo co-solvent formulations at ≥2.5 mg/mL.
  • Store protected from light at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.