Learn More
Medchemexpress LLC m-PEG7-amine | 170572-38-0 | MFCD13184965 | 98.0% | 339.43 g/mol | C15H33NO7 | 5 G
SDP
Catalog No. 5000202196
Encompass_Preferred
m-PEG7-Amine is a methoxy-terminated polyethylene glycol (PEG) linker bearing a terminal primary amine. It is used as a PROTAC linker and as a cleavable ADC linker for antibody-drug conjugation and other bioconjugation applications. The reagent provides a flexible, hydrophilic spacer suitable for linker chemistry in targeted degraders and conjugates.
  • Methoxy-terminated PEG with seven ethylene glycol units.
  • Terminal primary amine for facile coupling to carboxylates and activated esters.
  • Molecular weight 339.43 g/mol and CAS 170572-38-0.
  • Suitable for use as a PROTAC linker and a cleavable ADC linker.
  • Hydrophilic spacer that improves solubility and reduces aggregation in conjugates.
Catalog No. 50-002-02196 Supplier Medchemexpress LLC Supplier No. HY1202375G
m-PEG7-Amine is a methoxy-terminated polyethylene glycol (PEG) linker bearing a terminal primary amine. It is used as a PROTAC linker and as a cleavable ADC linker for antibody-drug conjugation and other bioconjugation applications. The reagent provides a flexible, hydrophilic spacer suitable for linker chemistry in targeted degraders and conjugates.
  • Methoxy-terminated PEG with seven ethylene glycol units.
  • Terminal primary amine for facile coupling to carboxylates and activated esters.
  • Molecular weight 339.43 g/mol and CAS 170572-38-0.
  • Suitable for use as a PROTAC linker and a cleavable ADC linker.
  • Hydrophilic spacer that improves solubility and reduces aggregation in conjugates.