Learn More
Medchemexpress LLC m-PEG7-CH2CH2COOH | 1093647-41-6 | 10 G
SDP
Catalog No. 5000340966
Encompass_Preferred
m-PEG7-CH2CH2COOH is a non-cleavable ADC linker utilized in the creation of antibody-drug conjugates (ADCs). It also serves as a PEG-based PROTAC linker for the synthesis of PROTACs. ADCs consist of an antibody linked to an ADC cytotoxin via an ADC linker. PROTACs feature two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other binds to the target protein. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Non-cleavable linker for antibody-drug conjugates
  • PEG-based linker for PROTACs
  • Exploits the ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
Catalog No. 50-003-40966 Supplier Medchemexpress LLC Supplier No. HY13015110G
m-PEG7-CH2CH2COOH is a non-cleavable ADC linker utilized in the creation of antibody-drug conjugates (ADCs). It also serves as a PEG-based PROTAC linker for the synthesis of PROTACs. ADCs consist of an antibody linked to an ADC cytotoxin via an ADC linker. PROTACs feature two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other binds to the target protein. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Non-cleavable linker for antibody-drug conjugates
  • PEG-based linker for PROTACs
  • Exploits the ubiquitin-proteasome system
  • Designed for selective degradation of target proteins