Learn More
Medchemexpress LLC m-PEG7-CH2CH2COOH | 1093647-41-6 | >99.39% | 25 G
SDP
Catalog No. 5000342081
Encompass_Preferred
m-PEG7-CH2CH2COOH (CAS No. 1093647-41-6) is a non-cleavable, PEG-based linker used in the synthesis of antibody-drug conjugates (ADCs) and as a PROTAC linker. It is an essential component for developing targeted therapeutics due to its role in connecting active pharmaceutical ingredients to targeting moieties.
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years.
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month.
  • Shipping at room temperature is acceptable if duration is less than 2 weeks.
  • Chemically identified as 4,7,10,13,16,19,22,25-Octaoxahexacosanoic acid.
Catalog No. 50-003-42081 Supplier Medchemexpress LLC Supplier No. 25GHY130151
m-PEG7-CH2CH2COOH (CAS No. 1093647-41-6) is a non-cleavable, PEG-based linker used in the synthesis of antibody-drug conjugates (ADCs) and as a PROTAC linker. It is an essential component for developing targeted therapeutics due to its role in connecting active pharmaceutical ingredients to targeting moieties.
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years.
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month.
  • Shipping at room temperature is acceptable if duration is less than 2 weeks.
  • Chemically identified as 4,7,10,13,16,19,22,25-Octaoxahexacosanoic acid.