Learn More
Medchemexpress LLC M-PEG7-CH2CH2COOH | 1093647-41-6 | 99.4% | 250 MG
SDP
Catalog No. 5000342712
Encompass_Preferred
M-PEG7-CH2CH2COOH is a non-cleavable ADC linker utilized in the creation of antibody-drug conjugates (ADCs). It also serves as a PEG-based PROTAC linker for the synthesis of PROTACs.
  • Used in antibody-drug conjugates (ADCs)
  • Functions as a PROTAC linker to selectively degrade target proteins via the ubiquitin-proteasome system
  • Non-cleavable linker
Catalog No. 50-003-42712 Supplier Medchemexpress LLC Supplier No. 250MGHY130151
M-PEG7-CH2CH2COOH is a non-cleavable ADC linker utilized in the creation of antibody-drug conjugates (ADCs). It also serves as a PEG-based PROTAC linker for the synthesis of PROTACs.
  • Used in antibody-drug conjugates (ADCs)
  • Functions as a PROTAC linker to selectively degrade target proteins via the ubiquitin-proteasome system
  • Non-cleavable linker