Learn More
Medchemexpress LLC m-PEG7-thiol | 651042-82-9 | 99.9% | 356.48 g/mol | C15H32O7S | 1 G
SDP
Catalog No. 5000209920
Encompass_Preferred
Thiol-terminated polyethylene glycol (PEG) linker suitable for use as a flexible, hydrophilic spacer in the synthesis of PROTACs and other bioconjugates. Provides a terminal thiol for selective thiol-reactive coupling and is supplied with supporting analytical and safety documentation.
  • Thiol-terminated PEG linker
  • Average molecular weight 356.48 g/mol
  • High purity: 99.9%
  • Available in 1 G and other pack sizes
  • Used for PROTAC synthesis and thiol-based conjugation
  • COA and SDS available for quality and safety information
Catalog No. 50-002-09920 Supplier Medchemexpress LLC Supplier No. HYW0520061G
Thiol-terminated polyethylene glycol (PEG) linker suitable for use as a flexible, hydrophilic spacer in the synthesis of PROTACs and other bioconjugates. Provides a terminal thiol for selective thiol-reactive coupling and is supplied with supporting analytical and safety documentation.
  • Thiol-terminated PEG linker
  • Average molecular weight 356.48 g/mol
  • High purity: 99.9%
  • Available in 1 G and other pack sizes
  • Used for PROTAC synthesis and thiol-based conjugation
  • COA and SDS available for quality and safety information