Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG8-amine | 869718-81-0 | MFCD13184961 | ≥98.0% | 383.48 g·mol⁻¹ | C17H37NO8 | 100 MG
SDP

Catalog No. 5000204089
Encompass_Preferred

m-PEG8-amine is an 8-unit monomethoxy polyethylene glycol (PEG) bearing a terminal primary amine used as a cleavable spacer/linker in antibody-drug conjugates and for general bioconjugation. It is supplied as a powder and is typically handled and stored according to the supplier's safety documentation.

  • Chemical name: m-PEG8-amine
  • Cas number: 869718-81-0
  • Purity: ≥98.0%
  • Molecular formula: C17H37NO8
  • Molecular weight: 383.48 g·mol⁻¹
  • Typical solubility: soluble in DMSO
  • Available pack sizes: 50 MG, 100 MG, 500 MG, 1 G, 5 G, 10 G, 50 G

Catalog No. 50-002-04089 Supplier Medchemexpress LLC Supplier No. HYW040236100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG8-amine is an 8-unit monomethoxy polyethylene glycol (PEG) bearing a terminal primary amine used as a cleavable spacer/linker in antibody-drug conjugates and for general bioconjugation. It is supplied as a powder and is typically handled and stored according to the supplier's safety documentation.

  • Chemical name: m-PEG8-amine
  • Cas number: 869718-81-0
  • Purity: ≥98.0%
  • Molecular formula: C17H37NO8
  • Molecular weight: 383.48 g·mol⁻¹
  • Typical solubility: soluble in DMSO
  • Available pack sizes: 50 MG, 100 MG, 500 MG, 1 G, 5 G, 10 G, 50 G

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.