Learn More
Medchemexpress LLC m-PEG8-azide | 869718-80-9 | 250 MG
SDP
Catalog No. 5000342491
Encompass_Preferred
m-PEG8-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent containing an Azide group, enabling specific reactions with other molecular groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Relevant for cancer research and targeted therapy applications
Catalog No. 50-003-42491 Supplier Medchemexpress LLC Supplier No. 250MGHY130204
m-PEG8-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent containing an Azide group, enabling specific reactions with other molecular groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Relevant for cancer research and targeted therapy applications