Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG8-CH2COOH | 102013-72-9 | 97.0% | 1 G
SDP

Catalog No. 5000340967
Encompass_Preferred

m-PEG8-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. It has a molecular weight of 442.50 and a molecular formula of C19H38O11. This compound is a colorless to light yellow liquid with a density of 1.122 g/cm3.

  • Used in the synthesis of PROTACs.
  • Functions as a PEG-based PROTAC linker.
  • Facilitates the degradation of target proteins by exploiting the ubiquitin-proteasome system.
  • Soluble in DMSO at 200 mg/mL (451.98 mM), with ultrasonic assistance potentially needed.
  • Molecular formula: C19H38O11.
  • Molecular weight: 442.50.
  • Target: PROTAC linkers.
  • Pathway: PROTAC.

Catalog No. 50-003-40967 Supplier Medchemexpress LLC Supplier No. HY1301451G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG8-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. It has a molecular weight of 442.50 and a molecular formula of C19H38O11. This compound is a colorless to light yellow liquid with a density of 1.122 g/cm3.

  • Used in the synthesis of PROTACs.
  • Functions as a PEG-based PROTAC linker.
  • Facilitates the degradation of target proteins by exploiting the ubiquitin-proteasome system.
  • Soluble in DMSO at 200 mg/mL (451.98 mM), with ultrasonic assistance potentially needed.
  • Molecular formula: C19H38O11.
  • Molecular weight: 442.50.
  • Target: PROTAC linkers.
  • Pathway: PROTAC.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.