Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG8-NHS ester | 756525-90-3 | MFCD13184969 | 95.0% | 509.54 g/mol | C22H39NO12 | 100 MG
SDP

Catalog No. 5000203219
Encompass_Preferred

m-PEG8-NHS ester is a methoxy polyethylene glycol (PEG) linker containing an N-hydroxysuccinimide (NHS) ester functional group. It provides an eight-unit PEG spacer for conjugation to primary amines and is used in antibody-drug conjugate (ADC) synthesis and general bioconjugation to increase solubility and reduce aggregation.

  • Non-cleavable 8-unit PEG spacer
  • Amine-reactive NHS ester for conjugation
  • Supplied as a 100 mg vial (research use only)
  • Purity by NMR: 95.0%
  • Molecular weight 509.54 g/mol; formula C22H39NO12
  • Store pure material at -20°C for long-term stability

Catalog No. 50-002-03219 Supplier Medchemexpress LLC Supplier No. HYW019793100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG8-NHS ester is a methoxy polyethylene glycol (PEG) linker containing an N-hydroxysuccinimide (NHS) ester functional group. It provides an eight-unit PEG spacer for conjugation to primary amines and is used in antibody-drug conjugate (ADC) synthesis and general bioconjugation to increase solubility and reduce aggregation.

  • Non-cleavable 8-unit PEG spacer
  • Amine-reactive NHS ester for conjugation
  • Supplied as a 100 mg vial (research use only)
  • Purity by NMR: 95.0%
  • Molecular weight 509.54 g/mol; formula C22H39NO12
  • Store pure material at -20°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.