Learn More
Medchemexpress LLC m-PEG8-thiol | 651042-83-0 | 99.9% | C17H36O8S | 100 MG
SDP
Catalog No. 5000354281
Encompass_Preferred
m-PEG8-thiol is a PEG-based PROTAC linker that facilitates the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs are bifunctional molecules comprising two ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein, thereby exploiting the intracellular ubiquitin-proteasome system for selective protein degradation.
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs for targeted protein degradation
  • Exploits the intracellular ubiquitin-proteasome system
Catalog No. 50-003-54281 Supplier Medchemexpress LLC Supplier No. HY141330100MG
m-PEG8-thiol is a PEG-based PROTAC linker that facilitates the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs are bifunctional molecules comprising two ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein, thereby exploiting the intracellular ubiquitin-proteasome system for selective protein degradation.
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs for targeted protein degradation
  • Exploits the intracellular ubiquitin-proteasome system