Learn More
Medchemexpress LLC m-PEG9-acid | 2576495-35-5 | MFCD27977514 | 98.0% | C20H40O11 | 250 MG
SDP
Catalog No. 5000352587
Encompass_Preferred
m-PEG9-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds consist of two ligands connected by a linker, one for an E3 ubiquitin ligase and the other for a target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system. This product is a colorless to light yellow liquid, with a molecular formula of C20H40O11 and a molecular weight of 456.53, intended for laboratory use only.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Enables selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
  • For laboratory and research use only
  • Colorless to light yellow liquid
  • Purity of 98.0%
Catalog No. 50-003-52587 Supplier Medchemexpress LLC Supplier No. 250MGHY140499
m-PEG9-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds consist of two ligands connected by a linker, one for an E3 ubiquitin ligase and the other for a target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system. This product is a colorless to light yellow liquid, with a molecular formula of C20H40O11 and a molecular weight of 456.53, intended for laboratory use only.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Enables selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
  • For laboratory and research use only
  • Colorless to light yellow liquid
  • Purity of 98.0%