Learn More
Medchemexpress LLC M-PEG9-amine | 211859-73-3 | 97.0% | 427.53 g/mol | C19H41NO9 | 100 MG
SDP
Catalog No. 5000202723
Encompass_Preferred
m-PEG9-amine is a methoxy-terminated polyethylene glycol (PEG) linker containing nine ethylene glycol units and a terminal primary amine. It is supplied as a liquid (colorless to light yellow) and is used as a PEG-based linker for proteolysis-targeting chimera (PROTAC) synthesis and as a cleavable linker in antibody-drug conjugate (ADC) development.
  • Methoxy-terminated polyethylene glycol (PEG) linker with a terminal primary amine.
  • Suitable for use as a PEG-based linker in PROTAC synthesis.
  • Functions as a cleavable linker for antibody-drug conjugates (ADCs).
  • High purity (97.0%) for research and conjugation workflows.
  • Molecular weight about 427.53 g/mol; formula C19H41NO9; CAS 211859-73-3.
  • Available in laboratory-scale sizes, including 100 mg, 250 mg, 1 g, and 5 g.
Catalog No. 50-002-02723 Supplier Medchemexpress LLC Supplier No. HY130571100MG
m-PEG9-amine is a methoxy-terminated polyethylene glycol (PEG) linker containing nine ethylene glycol units and a terminal primary amine. It is supplied as a liquid (colorless to light yellow) and is used as a PEG-based linker for proteolysis-targeting chimera (PROTAC) synthesis and as a cleavable linker in antibody-drug conjugate (ADC) development.
  • Methoxy-terminated polyethylene glycol (PEG) linker with a terminal primary amine.
  • Suitable for use as a PEG-based linker in PROTAC synthesis.
  • Functions as a cleavable linker for antibody-drug conjugates (ADCs).
  • High purity (97.0%) for research and conjugation workflows.
  • Molecular weight about 427.53 g/mol; formula C19H41NO9; CAS 211859-73-3.
  • Available in laboratory-scale sizes, including 100 mg, 250 mg, 1 g, and 5 g.