Learn More
Medchemexpress LLC m-PEG9-Amine | 211859-73-3 | >97.0% | 5 G
SDP
Catalog No. 5000340751
Encompass_Preferred
m-PEG9-Amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs, and also functions as a cleavable ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs). It facilitates the construction of compounds that either selectively degrade target proteins via the ubiquitin-proteasome system or enable targeted delivery of cytotoxic drugs.
  • Functions as a PEG-based PROTAC linker
  • Suitable for the synthesis of PROTACs
  • Acts as a cleavable ADC linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Supports targeted protein degradation via the ubiquitin-proteasome system
  • Enables targeted delivery of cytotoxic drugs
Catalog No. 50-003-40751 Supplier Medchemexpress LLC Supplier No. HY1305715G
m-PEG9-Amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs, and also functions as a cleavable ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs). It facilitates the construction of compounds that either selectively degrade target proteins via the ubiquitin-proteasome system or enable targeted delivery of cytotoxic drugs.
  • Functions as a PEG-based PROTAC linker
  • Suitable for the synthesis of PROTACs
  • Acts as a cleavable ADC linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Supports targeted protein degradation via the ubiquitin-proteasome system
  • Enables targeted delivery of cytotoxic drugs