Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Phenetidine, MP Biomedicals

Catalog No. ICN216479 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216479 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216479 Supplier MP Biomedicals, Inc Supplier No. 0521647905

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 621-33-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00007785
InChI Key WEZAHYDFZNTGKE-UHFFFAOYSA-N
Synonym m-phenetidine, benzenamine, 3-ethoxy, m-ethoxyaniline, 3-ethoxybenzenamine, unii-xe9kkl972r, ccris 4696, xe9kkl972r, 3-ethoxyphenylamine, 3-ethoxy aniline, 3-ethoxy-phenylamine
PubChem CID 12120
IUPAC Name 3-ethoxyaniline
SMILES CCOC1=CC=CC(N)=C1

Specifications

Quantity 5 g
Formula Weight 137.18
Chemical Name or Material M-Phenetidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.